Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MRTO4 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB087370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087370 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087370 Supplier Novus Biologicals Supplier No. NBP300348B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MRTO4 Polyclonal specifically detects MRTO4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MRTO4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias C1orf33, chromosome 1 open reading frame 33, dJ657E11.4, mRNA turnover 4 homolog (S. cerevisiae), mRNA turnover protein 4 homolog, MRT4, mRNA turnover 4, homolog, MRT4, mRNA turnover 4, homolog (S. cerevisiae), MRT460S acidic ribosomal protein PO
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant MRTO4 (Q9UKD2; Met1-Asp239).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51154
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.