Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A3 Antibody (489433), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB311783 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311783 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311783 Supplier Novus Biologicals Supplier No. FAB6247PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MS4A3 Monoclonal antibody specifically detects MS4A3 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen MS4A3
Applications Flow Cytometry
Classification Monoclonal
Clone 489433
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD20 antigen homolog, CD20 antigen-like protein, CD20LIgE receptor beta subunit, hematopoietic cell 4 transmembrane protein, Hematopoietic-specific transmembrane protein 4, HTm4, HTM4IgE receptor beta chain, membrane-spanning 4-domains subfamily A member 3, membrane-spanning 4-domains, subfamily A, member 3 (hematopoieticcell-specific)
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human MS4A3, Accession # NP_006129
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 932
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.