Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A4A Antibody (5C12), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB153772 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB153772 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB153772 Supplier Novus Biologicals Supplier No. NBP312043AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MS4A4A Monoclonal antibody specifically detects MS4A4A in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen MS4A4A
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 5C12
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 4SPAN1, CD20 antigen-like 1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4MGC22311, MS4A7
Host Species Rabbit
Immunogen This antibody was generated by immunizing C57BL/6 mice against mouse cell line 300-19 expressing transfected human membrane-spanning 4-domains subfamily A member 4A (MS4A4A)-GFP.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51338
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.