Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A4A Antibody (818112) [Biotin], Novus Biologicals™
SDP

Catalog No. NB374033 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374033 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374033 Supplier Novus Biologicals Supplier No. FAB7797B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MS4A4A Monoclonal specifically detects MS4A4A in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen MS4A4A
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 818112
Conjugate Biotin
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias 4SPAN1, CD20 antigen-like 1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4MGC22311, MS4A7
Host Species Mouse
Immunogen Mouse myeloma cell line NS0 transfected with human MS4A4A, Accession # Q96JQ5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 51338
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.