Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MS4A4A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP16000620UL
Description
MS4A4A Polyclonal specifically detects MS4A4A in Human samples. It is validated for Western Blot.Specifications
| MS4A4A | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| Q96JQ5 | |
| MS4A4A | |
| Synthetic peptides corresponding to MS4A4A(membrane-spanning 4-domains, subfamily A, member 4) The peptide sequence was selected from the N terminal of MS4A4A. Peptide sequence MHQTYSRHCRPEESTFSAAMTTMQGMEQAMPGAGPGVPQLGNMAVIHSHL. | |
| Affinity Purified | |
| RUO | |
| 51338 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| 4SPAN1, CD20 antigen-like 1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4MGC22311, MS4A7 | |
| Rabbit | |
| 26 kDa | |
| 20 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction