Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A4B Antibody (444008), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB369927 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369927 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369927 Supplier Novus Biologicals Supplier No. FAB4504Z
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MS4A4B Monoclonal specifically detects MS4A4B in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen MS4A4B
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 444008
Conjugate DyLight 755
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AI463180, Chandra, Ly116
Host Species Rat
Immunogen HEK293 human embryonic kidney cell line transfected with mouse MS4A4B, Met1-Val226, Accession # NP_068364
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60361
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.