Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A6A Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB185374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185374 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185374 Supplier Novus Biologicals Supplier No. NBP282106H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MS4A6A Polyclonal antibody specifically detects MS4A6A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen MS4A6A
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS without Preservative
Gene Alias 4SPAN3, CD20 antigen-like 3, CD20L3MS4A6A-polymorph, CD20-like precusor, Four-span transmembrane protein 3, four-span transmembrane protein 3.1,4SPAN3.2, four-span transmembrane protein 3.2, HAIRB-iso, membrane-spanning 4-domains subfamily A member 6A, membrane-spanning 4-domains, subfamily A, member 6A, MGC131944, MGC22650, MS4A6MST090, MSTP090
Host Species Rabbit
Immunogen MS4A6A antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human MS4A6A. The immunogen is located within the first 50 amino acids of MS4A6A. Amino Acid Squence: SQAEKPEPTNQGQDSLKKH
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64231
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.