Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MS4A8B Antibody, Novus Biologicals™
SDP

Catalog No. NB433981 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB433981 25ul
NBP181025 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB433981 Supplier Novus Biologicals Supplier No. NBP18102525UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MS4A8B Polyclonal specifically detects MS4A8B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MS4A8B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:10-1:20
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9BY19
Gene Alias 4SPAN4, Four-span transmembrane protein 4, membrane-spanning 4-domains subfamily A member 8B, membrane-spanning 4-domains, subfamily A, member 8B, MS4A4
Gene Symbols MS4A8
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:EFGIACASSHFGCQLVCCQSSNVSVIYPNIYAANPVITPEPVTSPPSYSSEIQANK
Purification Method Affinity Purified
Quantity 25ul
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83661
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.