Learn More
MSA™ Airline Respirator Manifolds
Catalog No. 18139753 Shop All MSA Products
Change view
Click to view available options
Connection Type:
Foster quick-disconnect, four-outlets
Foster quick-disconnect, two-outlets
Snap-Tite™ quick-disconnect, four-outlets
Snap-Tite™ quick-disconnect, two-outlets
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
18-139-753 Foster quick-disconnect, four-outlets
18-139-767 Foster quick-disconnect, two-outlets
18-139-760 Snap-Tite™ quick-disconnect, two-outlets
18-139-762 Snap-Tite™ quick-disconnect, four-outlets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-139-753 Supplier MSA™ Supplier No. 47370
Add to Cart
Add to Cart
Compressed airline manifolds help ensure proper air supply from a single air source.
  • Enhance long-duration respiratory protection
  • Ensure proper pressure and air quality
  • Regulator reduces air system compressor pressure and maintains desired operating pressure
  • Manifolds feature quick-disconnect assemblies and automatic shut-off outlets
Specifications
Connection Type Foster quick-disconnect, four-outlets
For Use With (Equipment) MSA Airline Respirators
Includes Automatic shut-off
Model 47370
Product Type Manifold Assembly