Description
- Enhance long-duration respiratory protection
- Ensure proper pressure and air quality
- Regulator reduces air system compressor pressure and maintains desired operating pressure
- Manifolds feature quick-disconnect assemblies and automatic shut-off outlets
Specifications
Specifications
| Connection Type | Snap-Tite™ quick-disconnect, two-outlets |
| For Use With (Equipment) | MSA Airline Respirators |
| Includes | Automatic shut-off |
| Model | 486053 |
| Product Type | Manifold Assembly |