Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ALTAIR™ 5 Multigas Detector Deluxe Kit

Catalog No. 18039322 Shop All MSA Products
Change view
Click to view available options
Battery Life:
15 hr.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Life
18-039-322 15 hr.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-039-322 Supplier MSA™ Supplier No. 10094917

missing translation for 'validMainframeMsg'

Includes

Altair 5 Multigas Detector with data logging, 1 ft. Probe, 10 ft. sampling line, integral pump, rechargeable battery pack, charger, calibration cap, tubing, CD manual

Featuring an exclusive end-of-sensor-life warning, this detector is Galaxy™ System-compatible for easy calibration and record keeping

  • Industry first 60-second span calibration and 10 second bump test
  • Thick rubber housing provides non-slip profile for secure grip, easily surviving 10ft drop
  • MotionAlert™ feature activates when a unit does not detect movement for 30 seconds
  • 40% less calibration gas used per minute than industry average
  • Sensor response and clear times in under 15 seconds on LEL, O2, CO, H2S, and SO2 — 50% faster than industry average
  • Greater signal stability and repeatability under changing or extreme
  • Six gas-capability

  • High-resolution color display screen
  • Many e-chem sensor options
  • Integral pump or diffusion option
  • 18-language display capability

Deluxe Kits — Color Display, 304.8cm (10 ft.) sampling line 30.48cm (1 ft.) probe

  • Chemical
  • Construction
  • Gas Detection
  • General Industry
  • Hazardous Materials
  • Indoor Air Quality
  • Manufacturing
  • Nuclear
  • Oil Gas
  • Telecommunications
  • Water/Wastewater

Certifications

  • CE ATEX I I 2G
  • Ex ia d IIC T4
  • UL Class I, Division 1, ABCDFG, Class II, Class III
  • CSA C22.2 No. 157 Class I, Div. 1, Groups ABCD
  • ANZ Ex ia s I/IIC T4 (Zone 0)
  • IEC Ex ia d e I/IIC T4

Warranty and Services

2 years under normal use conditions for LEL/O2/CO/H2S versions

Only gas meter in its class with internally-integrated pump

Specifications

Alarm Types Audible, visual, vibrating
Battery Life 15 hr.
Depth (English) 1.95 in.
Depth (Metric) 4.95 cm
Components Measured Carbon Monoxide, LEL, Oxygen, Phosphine
Dimensions (L x W x H) 6.68 x 3.52 x 6.68 in. (16.9 x 8.94 x 16cm)
Height (English) 6.68 in.
Height (Metric) 16 cm
Includes Altair 5 Multigas Detector with data logging, 1 ft. Probe, 10 ft. sampling line, integral pump, rechargeable battery pack, charger, calibration cap, tubing, CD manual
Length (English) 6.68 in.
Length (Metric) 16.9 cm
Resolution LEL: 1%, O2: 0.1%, CO: 1ppm, Phosphine 0.1ppm
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Weight (English) 18.4 oz.
Weight (Metric) 520g
Width (English) 3.52 in.
Width (Metric) 8.94 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.