Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ALTAIR™ 5 Multigas Detector Industrial Kit

Catalog No. 18039320 Shop All MSA Products
Change view
Click to view available options
Resolution:
LEL: 1%, O2: 0.1%, CO: 1ppm, Phosphine 0.1ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Resolution
18-039-320 LEL: 1%, O2: 0.1%, CO: 1ppm, Phosphine 0.1ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-039-320 Supplier MSA™ Supplier No. 10094907

missing translation for 'validMainframeMsg'

Includes

Altair 5X Multi-Gas Detector with Color Display, Lith-Ion Battery, 120V AC Charger, LEL,O2,CO,H2S Sensors, 304.8cm (10 ft.) sampling line, integral pump, calibration cap, 30.48cm (1 ft.) probe

Numerous customization options available, from personal monitoring and pre-entry checks within confined space to industrial hygienist use

  • Thick rubber housing provides a non-slip profile for secure grip, easily surviving a 6 ft. drop
  • MotionAlert™ feature activates when a unit does not detect movement for 30 seconds, indicating“man down”
  • InstantAlert™ feature offers manual activation of all alarms to alert others to potentially hazardous situations
  • Minimum IP65 rating for dust and water ingress
  • High resolution color display provides crystal clear visibility indoors or out, even in bright sunlight where lesser displays fail to provide critical information. Display may be customized using your company's logo
  • Monochrome display option provides cost management solutions without sacrificing basic functions expected from a high-end gas detector
  • Multi-language capability to serve your needs virtually anywhere in the world using one of many built-in languages
  • Galaxy™ System compatible for automatic testing, calibration and record keeping
  • MSA Link™ Software allows all stored records to be easily accessed for printed calibration reports, and alarm set point changes
  • Large, ergonomically designed buttons ensure easy reach and operation, even with gloved hands

  • Chemical
  • Construction
  • Gas Detection
  • General Industry
  • Hazardous Materials
  • Indoor Air Quality
  • Manufacturing
  • Nuclear
  • Oil Gas
  • Telecommunications
  • Water/Wastewater

Certifications

  • CE ATEX I I 2G
  • Ex ia d IIC T4
  • UL Class I, Division 1, ABCDFG, Class II, Class III
  • CSA C22.2 No. 157 Class I, Div. 1, Groups ABCD
  • ANZ Ex ia s I/IIC T4 (Zone 0)
  • IEC Ex ia d e I/IIC T4

Warranty and Services

2 years under normal use conditions for LEL/O2/CO/H2S versions

Specifications

Alarm Types Audible, visual, vibrating
Battery Life 15 hr.
Depth (English) 1.95 in.
Depth (Metric) 4.95 cm
Components Measured Carbon Monoxide, LEL, Oxygen, Phosphine
Dimensions (L x W x H) 6.68 x 3.52 x 6.68 in. (16.9 x 8.94 x 16cm)
Height (English) 6.68 in.
Height (Metric) 16 cm
Includes Altair 5X Multi-Gas Detector with Color Display, Lith-Ion Battery, 120V AC Charger, LEL,O2,CO,H2S Sensors, 304.8cm (10 ft.) sampling line, integral pump, calibration cap, 30.48cm (1 ft.) probe
Length (English) 6.68 in.
Length (Metric) 16.9 cm
Resolution LEL: 1%, O2: 0.1%, CO: 1ppm, Phosphine 0.1ppm
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Weight (English) 18.4 oz.
Weight (Metric) 520g
Width (English) 3.52 in.
Width (Metric) 8.94 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.