Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Bitrex™ Qualitative Fit-Test Kit

Catalog No. 19037721 Shop All MSA Products
Click to view available options
Product Type:
Replacement Fit Test Solution
Replacement Hood
Sensitivity Solution
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Two nebulizer bulbs, testing solution insert, sensitivity test solution insert, test hood, one replacement set of testing and sensitivity nebulizer inserts, and Using the Qualitative Fit-Test Kit videotape.

Safe, efficient, and reliable, this kit has everything necessary to perform OSHA-mandated fit testing on all NIOSH-approved air-purifying respirators.

Bitrex solution contains the chemical compound denatonium benzoate, making it an extremely bitter solution easily detected by the test subject.

Specifications

Product Type Sensitivity Solution
Capacity (English) 1.85 oz.
Capacity (Metric) 55 mL
Certifications/Compliance OSHA, NIOSH
For Use With (Equipment) MSA Bitrex and Saccharin qualitative fit test kit
Model 697445
Quantity 1 Ea.
Type (Solution) Sensitivity solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.