Description
Includes
Two nebulizer bulbs, testing solution insert, sensitivity test solution insert, test hood, one replacement set of testing and sensitivity nebulizer inserts, and Using the Qualitative Fit-Test Kit videotape.
Bitrex solution contains the chemical compound denatonium benzoate, making it an extremely bitter solution easily detected by the test subject.
Specifications
Specifications
| Product Type | Replacement Hood |
| Certifications/Compliance | OSHA, NIOSH |
| For Use With (Equipment) | MSA Bitrex and Saccharin qualitative fit test kit |
| Model | 697448 |
| Quantity | 1 Ea. |