Description
- Kits contains necessary calibration components to calibrate equipment
- Housed in lightweight carrying case for easy transport
Specifications
Specifications
| Description | For checking response of MSA gas sensors and gas detection instrumentation |
| Type | Calibration Kit |
| For Use With (Application) | Sensor and Instrument Calibration |
| Includes | Zero and span cylinders, regulators, tubing, accessories, instructions, and carrying case |