Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ESP™ II Voice Amplifier

Catalog No. 19003369 Shop All MSA Products
Click to view available options
:
ESP II voice amplifier

Hands-free voice amplification for local, short-range communication

Battery-powered, self-contained communications unit for use with the Millennium™ and Advantage™ 1000 facepieces

  • Provides hands-free voice amplification for local, short-range communication
  • Consists of a microphone and speaker
  • Housed within a waterproof, injection-molded, high-strength polymer case

Order Info

Weight: 8 oz. Batteries: two AAA, last up to 18 hours.

Certifications

NIOSH rated as intrinsically safe for use in Class I, II, and III, Division 1, Groups A, B, C, D, E, F, and G hazardous locations.

Specifications

Certifications/Compliance NIOSH
Product Type Voice Amplifier
For Use With (Application) Hands-free voice amplification for local, short-range communication
For Use With (Equipment) Millennium™ and advantage™ 1000 facepiece
Includes Microphone and speaker, push-button on/off
Model 10026265
Weight (English) 8 oz.
Weight (Metric) 0.226 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.