Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Fog-Reducing Nosecups

Catalog No. p-3492026 Shop All MSA Products
Click to view available options
Size:
Small
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

Color Black
Material Rubber
Product Type Nosecup Assembly
Size Small
For Use With (Application) Helps reduce lens fogging
Model 806466
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.