Description
- Single point of adjustment for easy donning and doffing
- Molds to body shape for comfort all day long
Fall Protection
Certifications
SEI Certified
Compliance
Meets ANSI Z359.1,ANSI A10.14, and OSHA requirements
Specifications
Specifications
| Color | Red |
| Material | Polyester |
| Style | Cross-Chest |
| Size | Standard |
| D-Ring Location | Back |
| Certifications/Compliance | Meets OSHA and ANSI requirements; CSA certified |
| Buckle Style | Qwik-Fit |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.