Learn More
MSA™ FP Pro™ Cross-Chest Style Harnesses
Catalog No. 19034688 Shop All MSA Products
Change view
Click to view available options
D-Ring Location:
Back
Back, Hips
Size:
Standard
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. D-Ring Location Size
19-034-688 Back X-Large
19-815-454 Back, Hips Standard
19-034-687 Back Standard
19-815-455 Back, Hips X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-034-688 Supplier MSA™ Supplier No. 415844
Pullover harness incorporating true sub-pelvic support for incredible comfort while suspending
  • Single point of adjustment for easy donning and doffing
  • Molds to body shape for comfort all day long
Fall Protection
Certifications
SEI Certified
Compliance
Meets ANSI Z359.1,ANSI A10.14, and OSHA requirements
Specifications
Color Red
Material Polyester
Style Cross-Chest
D-Ring Location Back
Buckle Style Qwik-Fit
Size X-Large
Certifications/Compliance Meets OSHA and ANSI requirements; CSA certified
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.