Learn More
MSA™ FP Pro™ Vest Style Harnesses
Catalog No. 19034690 Shop All MSA Products
Change view
Click to view available options
Buckle Style:
Qwik-Fit
Tongue
Color:
Red
D-Ring Location:
Back
Back, Hips
Size:
Standard
X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style Color D-Ring Location Size
19-034-690 Qwik-Fit Red Back Standard
19-042-271 Qwik-Fit Red Back, Hips X-Large
19-034-691 Qwik-Fit Red Back X-Large
19-999-331 Qwik-Fit Red Back, Hips Standard
19-130-3893 Tongue Red Back Standard
19-160-0188 Tongue Red Back, Hips X-Large
19-160-0189 Tongue Red Back, Hips Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-034-690 Supplier MSA™ Supplier No. 415947
Easy to doff and don, this harness features a full-body design offering excellent strength
  • Provides comfortable protection after long use in extreme conditions due to lightweight features and durable webbing
  • Integral belt and back pad for additional comfort and support for work positioning applications
  • Fits wide range of body types and sizes
  • Color-contrasting thigh and torso straps simplify donning
  • Bright red polyester webbing for high visibility
  • Capacity of 181.4kg (400 lb.)
Specifications
Color Red
Material Polyester
D-Ring Location Back
Style Vest
Buckle Style Qwik-Fit
Certifications/Compliance Meets OSHA and ANSI requirements; CSA certified
Size Standard
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.