Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ FP Pro™ Vest Style Harnesses

Catalog No. 191303893 Shop All MSA Products
Change view
Click to view available options
Buckle Style:
Qwik-Fit
Tongue
Color:
Red
D-Ring Location:
Back
Back, Hips
Size:
Standard
X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style Color D-Ring Location Size
19-130-3893 Tongue Red Back Standard
19-034-690 Qwik-Fit Red Back Standard
19-042-271 Qwik-Fit Red Back, Hips X-Large
19-034-691 Qwik-Fit Red Back X-Large
19-999-331 Qwik-Fit Red Back, Hips Standard
19-160-0188 Tongue Red Back, Hips X-Large
19-160-0189 Tongue Red Back, Hips Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-130-3893 Supplier MSA™ Supplier No. 10033836

Easy to doff and don, this harness features a full-body design offering excellent strength

  • Provides comfortable protection after long use in extreme conditions due to lightweight features and durable webbing
  • Integral belt and back pad for additional comfort and support for work positioning applications
  • Fits wide range of body types and sizes
  • Color-contrasting thigh and torso straps simplify donning
  • Bright red polyester webbing for high visibility
  • Capacity of 181.4kg (400 lb.)

Specifications

Color Red
Material Polyester
Buckle Style Tongue
Style Vest
D-Ring Location Back
Certifications/Compliance Meets OSHA and ANSI requirements; CSA certified
Size Standard
For Use With (Application) For fall arrest applications

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.