Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Gravity™ Harnesses

Catalog No. 19150956 Shop All MSA Products
Change view
Click to view available options
Material:
Nylon
Polyester
Size:
Standard
X-Large
Style:
Cross-over
Vest
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Size Style
19-150-956 Nylon X-Large Cross-over
19-160-0192 Nylon Standard Vest
19-150-954 Nylon Standard Cross-over
19-160-0190 Polyester Standard Vest
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-150-956 Supplier MSA™ Supplier No. 10067415

Curvilinear Comfort System provides total comfort during use

For controlled descent, positioning, ladder climbing, rescue and fall arrest.

  • Integral belt and comfortable saddle assist with support, position, restraint and climb movements
  • PFTE fabric protected webbing resists water, oil, grease and dirt
  • Capacity rating: 180kg (400 lb.)
  • Load-indicator alerts when the harness has been involved in a fall event
  • Standard size; adjustable

Certifications

CSA certified; Certified according EN 361

Specifications

Color Red, Blue
Material Nylon
Certifications/Compliance CSA certified; Certified according EN 361
For Use With (Application) Ladder climbing, controlled descent, rescue, fall arrest
Size X-Large
D-Ring Location Back, Front, Shoulder, Side
Style Cross-over

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.