Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ HAZMATCAD™ Hazardous Material Chemical Agent Detectors

Catalog No. p-4909671 Shop All MSA Products
Change view
Click to view available options
Type:
HAZMATCAD Detector for Nerve Gas
HAZMATCAD Plus Gas Detector
Includes:
Detector only
Detector, Blister Agents
Detector, Blister Agents, HCN
Detector, Blister Agents, phosgene
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Includes
19-130-1090 HAZMATCAD Detector for Nerve Gas Detector, Blister Agents
19-130-1091 HAZMATCAD Detector for Nerve Gas Detector, Blister Agents, HCN
19-130-1092 HAZMATCAD Detector for Nerve Gas Detector, Blister Agents, phosgene
19-130-1093 HAZMATCAD Plus Gas Detector Detector only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-130-1090 Supplier MSA™ Supplier No. 10055094

Easy-to-use, reliable detectors for first responders

Lightweight, portable detectors screen and classify a broad range of chemical threats including chemical warfare agents and selected toxic industrial chemicals (TICs). These detectors utilize miniaturized Surface Acoustic Wave (SAW) technology with innovative sample collector to display and alarm for presence of hydrogen cyanide, nerve gas, blister agents, or phosgene.

•    Rugged, compact and lightweight construction with special inlet design to protect against dust and particulate
•    Simple, intuitive user interface is easy to use without prior training, allowing rapid field deployment and operation in a level A environment
•    A single key press begins automatic sampling and analysis
•    Audible and visual alarm messages provide clear and simple alarm condition display
•    Offers self-diagnostic check during rapid warm-up, alphanumeric display with LED alarms and 8-12 hour operation with rechargeable lithium ion batteries
•    Vapor diffusion check source verifies system performance
•    RS 232 and IrDA communication ports store up to eight hours of data
•    HAZMATCAD Plus unit adds electrochemical sensor arrays to detect an additional four classes of toxic industrial chemicals (phosgene, hydrogen cyanide, halogen and hydride gases)
Dimensions: 2.3 × 7.9 × 9.8 in. (5.8 × 20 × 24.9cm). Weight (with batteries): 50 oz. (1.4kg).

Order Info

Both detectors supplied with RS-232 data cable, software, 8-hour datalogger, belt clip arm band, rubber boot, two long-life lithium ion rechargeable batteries, battery charger, vapor check source, a waterproof carrying case, one-year warranty, and user manual.

Specifications

Alarm Types Audible
Battery Life 8 to 12 hr.
Dimensions (L x W x H) 2.3 x 2.5 x 7.8 in. (5.84 x 6.35 x 19.81cm)
Type HAZMATCAD Detector for Nerve Gas
For Use With (Application) Screen and classify a broad range of chemical threats including chemical warfare agents and selected toxic industrial chemicals (TICs)
Height (English) 7.8 in.
Height (Metric) 19.81 cm
Includes Detector, Blister Agents
Length (English) 2.3 in.
Length (Metric) 5.84 cm
Temperature Range (English) 32°F to 105°F
Temperature Range (Metric) 0°C to 40°C
Warranty 1 Year
Width (English) 2.5 in.
Width (Metric) 6.35 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.