Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSA™ HAZMATCAD™ Hazardous Material Chemical Agent Detectors: Instruction Manuals
For HAZMATCAD and HAZMATCAD Plus Hazardous Material Chemical Agent Detectors.
Specifications
Alarm Types | Audible, visual |
---|---|
Battery Life | 8 to 12 hr. |
Type | Instruction Manual |
Warranty | 1 Year |
Weight (English) | 49.28 oz. |
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
19-130-1101
|
MSA™
10055753 |
MSA HAZMATCAD Plus Hazardous Material Chemical Agent Detector | N/A | N/A | N/A | ||||
19-130-1100
|
MSA™
10055752 |
MSA HAZMATCAD Hazardous Material Chemical Agent Detector | N/A | N/A | N/A | ||||
Description
For HAZMATCAD and HAZMATCAD Plus Hazardous Material Chemical Agent Detectors.Specifications
Audible, visual | |
Instruction Manual | |
49.28 oz. |
8 to 12 hr. | |
1 Year | |
1400g |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title