Learn More
MSA™ Passport™ Personal Alarms
Catalog No. 189994695 Shop All MSA Products
Change view
Click to view available options
3 Gas
4 Gas
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-999-4695 3 Gas
18-999-4696 4 Gas
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-4695 Supplier MSA™ Supplier No. 803892
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
Kits listed in the ordering table include sensors for CO, O2, LEL, and (for Cat. No. 18-999-4696) H2S; add-on pump module; standard NiCd battery pack; 120VAC charger; 10 ft. (3m) sampling line; 12 in. (30.5cm) probe tube; earphone, operating and technical manuals; and convenient instruction card in a cardboard carrying case.
Monitor and display multiple gas readings simultaneously. Ideal for monitoring combustible gases, oxygen, and toxic gases in workplace atmospheres, especially confined spaces. Built for high performance, the metal-impregnated polycarbonate case provides protection from electromagnetic interference (EMI) and radio-frequency interference (RFI). Menu-driven for ease of operation. Wide range of measurement options, including peak exposure, short-term exposure limit (STEL) and time-weighted average (TWA). Also provides readings on battery voltage and condition. Simplified calibration eliminates need to open instrument. Internal sensors monitor gases by diffusion or by remote sampling using pump module and sampling equipment. Available with three- and four-gas sensors. Combustible-gas sensor has range of 0 to 100% lower explosive limit (LEL); oxygen sensor, 0 to 25% by volume; carbon monoxide sensor, 0 to 2000ppm; and hydrogen sulfide sensor, 0 to 200ppm. Each monitor features three pushbuttons for simple operation: ON/OFF (activates unit), PAGE (accesses available display pages), and RESET (turns off alarm). For dark work areas, a backlight illuminates display when any button is activated. Measures combustible gas as a percentage of LEL or over a range of 0 to 5% methane; oxygen, a percentage by volume; and toxic gas in parts per million. Features exclusive Lockalarm™ Circuit that is “LEL-latching” for combustible and toxic gases. If atmospheric conditions reach 100% LEL, the alarm sounds and unit must be moved into a fresh-air environment to be silenced. Alarm setpoint is adjustable for each individual gas. Each monitor measures 8 1/8L × 1 7/8W × 3 1/4 in.H (20.6 × 4.8 × 8.3cm) and weighs 33 oz. (0.94kg).
Compliance
Passport Personal Alarms are MET approved for use in Class I, Division 1, Groups A, B, C, and D; Class II, Division 1, Groups E, F, and G; and Class III hazardous locations. Also CSA approved for Class I, Division 1, Groups A, B, C, and D locations.