Learn More
MSA™ Pressure Demand Air-Line Respirators
Shop All MSA ProductsDescription
Full-facepiece respirators feature belt-mounted regulator that maintains slight positive pressure of air inside facepiece during inhalation and exhalation to prevent influx of contaminants. Assembly includes regulator and quick-disconnect, breathing tube, Ultravue™ facepiece, and web belt. Facepiece features scratch-resistant polycarbonate lens with wraparound design for minimal visual distortion, inhalation check valve that prevents exhaled air from entering breathing tube or regulator, mechanical speaking diaphragm for short-range communications, and five adjustable rubber straps and a vinyl neck strap for holding facepiece in standby position.
Respirators are designed for use in non-IDLH atmospheres and are suitable for applications such as welding and painting, and for handling certain hazardous materials. Available with stainless-steel or Foster steel quick-disconnect.
Compliance
NIOSH/MSHA approved for operation at 65 to 85psig using 8 to 300 ft. (2.4 to 91.4m) of MSA airline hose.
Specifications
Specifications
| Certifications/Compliance | NIOSH, MSHA |
| Material | Foster Steel |
| Product Type | Quick-Disconnect |
| For Use With (Application) | Non-IDLH atmospheres, welding and painting and for handling certain hazardous materials |
| Includes | Regulator and quick-disconnect, breathing tube, ultravue™ facepiece and web belt |
| Material (Lens) | Polycarbonate |
| Pressure | 65 to 85 psi |
| Resistant To | Scratch-resistant |
| Length (English) Hose | 8 to 300 ft. |
| Length (Metric) Hose | 2.43 to 91.44 m |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.