Learn More
Specifications
Specifications
| Certifications/Compliance | NIOSH 42CFR, part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002 |
| Color | Black |
| Product Type | MMR Facepiece |
| For Use With (Application) | For monitoring air cylinder contents, firefighting |
| For Use With (Equipment) | Ultralite MMR and custom 4500 MMR air mask |
| Includes | Quarter turn quick-connect adapter, rubber head harness and nosecup |
| Material | Silicone |
| Size | Large |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.