Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSA™ Safe Escape CBRN Respirators
Shop All MSA Products
Click to view available options
Product Type:
Standard Escape Respirator
Tactical Escape Respirator
Size:
Small
Medium
Large
Description
- Translucent hood with one-piece large bonded lens offers secure protection and a wide field of view
- Canister is permanently bonded to the respirator for a secure connection
- Rubber neck seal provides a comfortable and reliable seal
- Elastic hair tie allows for the securing of hair before donning the respirator
- Nose cup helps reduce lens fogging and carbon dioxide buildup
- Hexagonal storage case is strong, lightweight, and easily accessible
- 15 minute air supply
Standard
- Color-coded labels on respirator
- Black/gray labels on respirator
Guards against HD vapor, GB, Ammonia, Cyanogen chloride, Cyclohexane, Formaldehyde, Hydrogen cyanide, Hydrogen sulfide, Nitrogen dioxide, Phosgene, Phosphine, sulfur dioxide
Certifications
NIOSH - 10052317
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Product Type | Tactical Escape Respirator |
| Duration | 15 min. |
| Cycle Time | 15 min. |
| For Use With (Application) | CBRN incidents, civil disturbances, clandestine labs |
| Includes | Bonded lens and cbrn canister, high stretch rubber neck seal, nose cup |
| Model | 10066788 |
| Size | Medium |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction