Learn More
MSA™ Sirius™ Multigas Detector - Economy Model
Combines PID functionality with a four-gas monitor
Supplier: MSA™ 10051152
Includes: Economy Models include: Monitor, sensors and batteries
Description
Integrates high-performance photoionization detector sensor with a multigas detector for up to four gases. Users are able to simultaneously monitor for volatile organic compounds (VOCs) with low vapor pressures while measuring for combustible, toxic, and oxygen-deficient atmospheres within one reliable, easy-to-use, durable unit.
Features:
• Measures VOC readings first for greater accuracy of ±10% or 2ppm over full range
• Best in class humidity performance
• Notifies failure or removal of lamp or ion chamber
• Features an over range indicator, external exhaust, and multidirectional audio and visual alarms
• Lamps are easy to replace and clean
For use in:
• Leak detection
• Perimeter monitor
• Spill delineation
• Remediation
• Arson investigation
• Diesel fuel TVL monitoring (Marine Chemist)
• Decontamination
• PPE Determination
Compliance
Meets 47CFR part 15. UL913 6th edition and CSA 22.2 No. 157 for use in hazardous locations Class I, Divisions I, Groups A, B, C, and D. Temperature class T3C. Meets Canadian combustible performance standard CSA 22.2 No. 152 for methane only.
Specifications
| 6.65 cm | |
| Monitor, Sensors and Batteries | |
| 47CFR, UL913, CSA 22.2, T3C |
| 0 to 100% LEL, 0 to 25% Vol O2, 0 to 200 ppm H2S | |
| ±10% | |
| 16.51 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.