Learn More
MSA™ Tee Blocks and Pigtails
Catalog No. p-4396282 Shop All MSA Products
Click to view available options
Connection Type:
Air pigtail
Air tee block
Material:
Brass
Tubular Copper
Product Type:
Pigtail
Tee Block
Tee blocks and pigtails are used to connect a bank of cylinders together.
Used to connect a bank of cylinders together
  • Coupler tees make a connection between the supply cylinder valve and the pigtail
  • Outlet connection are constructed of brass, while the pigtails joining the supply cylinders are made of tubular copper
  • 27 in. long air pigtail
Specifications
Connection Type Air tee block
For Use With (Equipment) Compressed Airline Respirator
Material Brass
Model 68850
Pressure 3000 psi
Product Type Tee Block
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.