Learn More
MSA™ Ultra Elite™ M7 Facepiece
Catalog No. 17200021 Shop All MSA Products
Click to view available options
Product Type:
Facepiece Assembly
M-7 Facepiece
Size:
Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Speed-on™ head harness, nosecup, firehawk™ m7 heads up display bracket, standard speaking diaphragm
For use with FireHawk™ M7 air masks
Provides enhanced peripheral and downward vision
  • Resists chemical attack, temperature extremes, and rugged use without tearing
  • Seals securely with wide range of facial contours
  • Five adjustable straps with stainless-steel, field-replaceable buckles for comfortable fit
  • Standard-speaking diaphragm
  • Rugged polycarbonate lens with scratch-resistant coating
  • Choice of push-to-connect or slide-to-connect style regulators
Specifications
Certifications/Compliance NFPA 2007
Product Type Facepiece Assembly
For Use With (Application) Resists chemical attack, temperature extremes, rugged use without tearing
For Use With (Equipment) Firehawk™ m7 air mask
Includes Speed-on™ head harness, nosecup, Firehawk™ M7 heads-up display bracket, standard speaking diaphragm
Size Large
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.