Learn More
MSA™ V-Gard™ 500 Vented Protective Cap
Shop All MSA Products
Click to view available options
Color:
Blue
Green
Hi-vis Orange
Red
White
Suspension Type:
4-Point Ratchet
6-Point Ratchet
Description
- Comfortable, lightweight design
- Polyethylene shell features contours above the ear for easy integration of hearing protection
- Glaregard™ underbrim reduces reflective glare
- Available with four- or six-point Fas-Trac™ ratchet suspension with absorbent sweatband
Compliance
Third-party certified to meet ANSI/ISEA Z98.1, Type I, Class C requirements.
Specifications
Specifications
| Certifications/Compliance | Meets or exceeds ANSI Z89.1-2003, Class C and CSA Z94.1-2005. |
| Color | Green |
| Suspension Type | 4-Point Ratchet |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.