Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ V-Gard™ Chin Protectors

Catalog No. p-4512030 Shop All MSA Products
Click to view available options
Product Type:
Retractable Chin Protector
Standard Chin Protector
Height (English):
2.86 in.
3.82 in.
Height (Metric):
7.2 cm
9.7 cm

Provide extra protection from impact and splash hazards to face and neck

  • For use with select V-Gard Nitrometers
  • Opaque material ensures safety and standards compliance with clear, tinted, shaded and coated V-Gard visors
  • Compact, lightweight standard protector provides additional protection between and neck area
  • Retractable protector allows greater flexibility of head movement

Specifications

Certifications/Compliance ANSI Z87.1 2010-IAC-03-005-CAL
Color Black
For Use With (Equipment) V-Gard Nitrometer Visor
Height (English) 3.82 in.
Height (Metric) 9.7 cm
Material Polycarbonate
Product Type Retractable Chin Protector
Style Opaque compact
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.