Learn More
MSA™ V-Gard™ Protective Slotted Cap with Staz-On™ Suspension
Catalog No. 189993985 Shop All MSA Products
Click to view available options
Color:
Black
Blue
Green
Hi-vis Orange
Orange
Pink
Purple
Red
White
Yellow
Size:
Standard
Lightweight for all-day wearing comfort
  • Four-point webbed Staz-On suspension with perforated vinyl sweatband
  • Sizes: Small (6-7 1/8), Standard (6 1/2-8), and Large (7-8 1/2)
  • Available in more than 20 colors, with custom imprinting and striping available
Order Info
Contact your Fisher Safety Customer Safety Representative for ordering information for sizes and colors not shown.
†With lamp bracket and cord holder (Class C)
Specifications
Certifications/Compliance Meets or exceeds ANSI Z89.1-2009; Type I; Class E; SEI certified.
Color Black
Size Standard
Suspension Type Staz-On Pin-Lock
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.