Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ XCell Sensor Kits

Catalog No. 17200132 Shop All MSA Products
Change view
Click to view available options
Components Measured:
Ammonia
Carbon Monoxide, Hydrogen Sulfide
Chlorine
Oxygen
Sulfur Dioxide
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
17-200-132 Carbon Monoxide, Hydrogen Sulfide
17-200-130 -
17-200-131 Oxygen
17-200-134 Chlorine
17-200-136 Sulfur Dioxide
17-200-138 Ammonia
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-200-132 Supplier MSA™ Supplier No. 10106725
Add to Cart
Add to Cart

Specifications

Components Measured Carbon Monoxide, Hydrogen Sulfide
For Use With (Equipment) Altair 5X Multigas Detector
Type Replacement Sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.