Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH2 Antibody (MSH2/6549R), Biotin, Novus Biologicals™
SDP

Catalog No. NB389046 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389046 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389046 Supplier Novus Biologicals Supplier No. NBP308958B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MSH2 Monoclonal antibody specifically detects MSH2 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen MSH2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MSH2/6549R
Conjugate Biotin
Formulation PBS
Gene Alias COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2
Host Species Rabbit
Immunogen Recombinant full-length human MSH2 protein (Uniprot: IDP43246)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4436
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.