Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH2 Mouse anti-Human, Clone: 25D12, Novus Biologicals™
SDP

Catalog No. NBP226489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226489 0.25 mL
NB019411 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP226489 Supplier Novus Biologicals Supplier No. NBP226489

Mouse Monoclonal Antibody

MSH2 Monoclonal specifically detects MSH2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen MSH2
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 25D12
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:10-1:25, Immunohistochemistry-Frozen 1:10 - 1:2000
Formulation Tissue culture supernatant with 0.05% Sodium Azide
Gene Alias COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2
Gene Symbols MSH2
Host Species Mouse
Immunogen BALB/C mice were injected with recombinant human MSH2 protein.
Purification Method Tissue culture supernatant
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4436
Test Specificity This antibody reacts with a 102 kDa protein known as MSH2 (mismatch repair protein 2).
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.