Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH3 Antibody (CL14064), Novus Biologicals™
SDP

Catalog No. NB305993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305993 25 μL
NB305994 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB305993 Supplier Novus Biologicals Supplier No. NBP32458825UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MSH3 Monoclonal antibody specifically detects MSH3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MSH3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL14064
Conjugate Unconjugated
Dilution Western Blot 1ug/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 2-10 ug/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias Divergent upstream protein, DNA mismatch repair protein Msh3, DUC1, DUG, DUPMGC163306, hMSH3, Mismatch repair protein 1, MRP1MGC163308, mutS (E. coli) homolog 3, mutS homolog 3 (E. coli)
Host Species Mouse
Immunogen This antibody was designed to target the recombinant protein MSH3, with the precise immunogen sequence being withheld for proprietary reasons.
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline DNA Repair, Mismatch Repair, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4437
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.