Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH5 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB318837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318837 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318837 Supplier Novus Biologicals Supplier No. NBP298819MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MSH5 Polyclonal antibody specifically detects MSH5 in Human samples. It is validated for Western Blot

Specifications

Antigen MSH5
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp434C1615, G7, hMSH5, MGC2939, mutS (E. coli) homolog 5, mutS homolog 5 (E. coli), mutS protein homolog 5, MUTSH5, NG23
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human MSH5.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Mismatch Repair, modENCODE Antibodies
Primary or Secondary Primary
Gene ID (Entrez) 4439
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.