Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH6 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292163 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292163 Supplier Novus Biologicals Supplier No. NB100328APC
Add to Cart
Add to Cart

Goat Polyclonal Antibody

MSH6 Polyclonal antibody specifically detects MSH6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation (Negative)

Specifications

Antigen MSH6
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein
Host Species Goat
Immunogen Maps to a region between residues 350 and 400 of human MutS homolog 6 using the numbering given in entry NP_000170.1 (GeneID 2956).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Repair, Mismatch Repair
Primary or Secondary Primary
Gene ID (Entrez) 2956
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.