Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSH6 Antibody (MSH6/3086), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB010966 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010966 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010966 Supplier Novus Biologicals Supplier No. NBP279846C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MSH6 Monoclonal specifically detects MSH6 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MSH6
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MSH6/3086
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein
Gene Symbols MSH6
Host Species Mouse
Immunogen Recombinant fragment of human MSH6 protein (around aa 374-540) (exact sequence is proprietary) (Uniprot: P52701)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Repair, Mismatch Repair
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.