Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSK1/RPS6KA5 Antibody (PCRP-RPS6KA5-1A8), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB382049 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382049 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382049 Supplier Novus Biologicals Supplier No. NBP308282AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MSK1/RPS6KA5 Monoclonal antibody specifically detects MSK1/RPS6KA5 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen MSK1/RPS6KA5
Applications Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-RPS6KA5-1A8
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, MGC1911,90 kDa ribosomal protein S6 kinase 5, MSK1EC 2.7.11.1, MSPK1, Nuclear mitogen- and stress-activated protein kinase 1, ribosomal protein S6 kinase alpha-5, ribosomal protein S6 kinase, 90kD, polypeptide 5, ribosomal protein S6 kinase, 90kDa, polypeptide 5, RLPK, RSKL, RSK-like protein kinase, S6K-alpha-5
Host Species Mouse
Immunogen Recombinant full-length human MSK1/RPS6KA5 protein (Uniprot: O75582)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9252
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.