Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSP/MST1 Antibody (OTI1A10), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272792A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272792A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272792A405 Supplier Novus Biologicals Supplier No. NBP272792AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MSP/MST1 Monoclonal specifically detects MSP/MST1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MSP/MST1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1A10
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias D3F15S2, EC 3.4.21, hepatocyte growth factor-like protein homolog, HGFLhepatocyte growth factor-like protein, macrophage stimulating 1 (hepatocyte growth factor-like), Macrophage stimulatory protein, Macrophage-stimulating protein, MSPDNF15S2, NF15S2
Gene Symbols MST1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 194-462 of human MSP/MST1 (NP_066278) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4485.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.