Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSRA Monoclonal Antibody (C6), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $458.50
Specifications
Antigen | MSRA |
---|---|
Clone | C6 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA550974
|
Invitrogen™
MA550974 |
20 μL |
Each for $195.80
|
|
|||||
FEMA550973
|
Invitrogen™
MA550973 |
200 μL |
Each for $458.50
|
|
|||||
Description
Methionine sulfoxide reductase (MsrA) reduces methionine sulfoxide (MetO) residues in proteins and free MetO to Methionine (Met). The catalytic activity of MsrA is dependent of bound metal and cofactors but it requires reducing equivalents from either DTT or a thioredoxin-regenerating system. MsrA plays an essential role in protecting cells against oxidative damage. The substrates of MsrA include calmodulin, HIV protease and a 1-proteinase-inhibitor. Recent studies indicate that there is a connection between MsrA and Alzheimer's disease in mammals.Specifications
MSRA | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
2310045J23Rik; 6530413P12Rik; cytosolic methionine-S-sulfoxide reductase; methionine sulfoxide reductase A; mitochondrial peptide methionine sulfoxide reductase; Msra; MSR-A; peptide met (O) reductase; peptide Met(O) reductase; peptide methionine sulfoxide reductase; peptide-methionine (S)-S-oxide reductase; PMSR; protein-methionine-S-oxide reductase | |
Msra | |
IgG1 κ | |
Protein A/G | |
Msra |
C6 | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat, Pig | |
Q923M1, Q9UJ68 | |
100152960, 29447, 4482 | |
Recombinant Human MSRA protein, Ala27-Lys235 (Accession #Q9UJ68). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title