Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSRB2 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB049998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB049998 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB049998 Supplier Novus Biologicals Supplier No. NBP297958V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MSRB2 Polyclonal specifically detects MSRB2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MSRB2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CBS1, CBS-1methionine-r-sulfoxide reductase B2 variant 3, CGI-131, EC 1.8.4, EC 1.8.4.-, EC 1.8.4.12, methionine sulfoxide reductase B, methionine sulfoxide reductase B2, methionine-r-sulfoxide reductase B2 variant 1, methionine-R-sulfoxide reductase B2, mitochondrial, MGC26104, MSRB, MsrB2, PILB, pilin-like transcription factor
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MSRB2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22921
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.