Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MST1/STK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP30366820UL

 View more versions of this product

Catalog No. NB092612


Only null left
Add to Cart

Description

Description

MST1/STK4 Polyclonal specifically detects MST1/STK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications

Specifications

MST1/STK4
Polyclonal
PBS with 50% glycerol, pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 303-487 of human MST1/STK4 (NP_006273.1).
20 μL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown
Unconjugated
Rabbit
Affinity purified
RUO
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.