Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MST1/STK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$220.00
Specifications
Antigen | MST1/STK4 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
MST1/STK4 Polyclonal specifically detects MST1/STK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
MST1/STK4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, KnockDown | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Recombinant fusion protein containing a sequence corresponding to amino acids 303-487 of human MST1/STK4 (NP_006273.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title