Learn More
Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent
Description
Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.
Use Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent for GC analyses of early eluting compounds that would otherwise be obscured in the chromatogram. MSTFA, the most volatile TMS-amide available, and its byproduct N-methyltrifluoroacetamide typically elute with the solvent front, allowing TMS derivatives of small molecules to be analyzed. Addition of Thermo Scientific™ TMCS aids derivatization of amides, secondary amines, and hindered hydroxyls not derivatized by MSTFA alone.
Specifications
Specifications
| For Use With | GC/MS |
| Standard Concentration | 98 to 100% |
| Hazards | Flammable, Irritant |
| Quantity | 100 mL |
| Standard Components | MTBSTFA |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.