Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent
GSA_VA

Catalog No. pi48914
Encompass
Click to view available options
Standard Components:
MSTFA
MSTFA + 1% TMCS
MTBSTFA
Quantity:
10 g
10 x 1 mL ampules
100 mL
25 mL

Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.

Use Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent for GC analyses of early eluting compounds that would otherwise be obscured in the chromatogram. MSTFA, the most volatile TMS-amide available, and its byproduct N-methyltrifluoroacetamide typically elute with the solvent front, allowing TMS derivatives of small molecules to be analyzed. Addition of Thermo Scientific™ TMCS aids derivatization of amides, secondary amines, and hindered hydroxyls not derivatized by MSTFA alone.

  • Trimethylsilyl donor strength comparable to BSA and BSTFA
  • Reacts to replace labile hydrogens on a wide range of polar compounds with a Si(CH3)3 group
  • Used to prepare volatile and thermally stable derivatives for GC and MS
  • Volatile byproduct, N-methyltrifluoroacetamide, has an even lower retention time than MSTFA
  • Often TMS derivatives of small molecules can be analyzed when derivatized with MSTFA because the byproducts and the reagent itself usually elute with the solvent front
  • Addition of Thermo Scientfic TMCS aids derivatization of amides, secondary amines and hindered hydroxyls not derivatized by MSTFA alone

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With GC/MS
Hazards Flammable, Irritant
Standard Concentration 98 to 100%
Storage 2°C to 8°C
Quantity 100 mL
Standard Components MSTFA

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.