Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSX1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB105470 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105470 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105470 Supplier Novus Biologicals Supplier No. NBP298746AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MSX1 Polyclonal specifically detects MSX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MSX1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias Homeobox protein Hox-7, homeobox protein MSX-1, HOX7homeobox 7, HYD1msh homeobox homolog 1 (Drosophila), msh (Drosophila) homeo box homolog 1 (formerly homeo box 7), msh homeo box 1, msh homeobox 1, Msh homeobox 1-like protein, msh homeobox homolog 1, OFC5, STHAG1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human MSX1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4487
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.